NanoDrop 3300
NanoDrop 3300 Fluorospectrometer
Fluorescence Made Compact and Simple
Thermo Scientific NanoDrop 3300 fluorospectrometers are used by life science laboratories to verify selective concentrations of DNA, RNA, and protein. Utilizing a patented sample retention system it minimizes the mass detection limit while increasing sensitivity, allowing measurement of samples as low as 1 picogram/µl of dsDNA. The flexibility of the NanoDrop 3300 allows users to work with multiple brands of assays, giving them the freedom to select the best assay for a specific workflow.
NanoDrop 3300 Fluorospectrometer features:
- Measures a wide variety of fluorescence assays: RiboGreen, PicoGreen, GFP, BFP mutants, Hoechst, 4-MU, Quantum Dots, OPA, DyLite, AlexaFluor and many more
- Sample size as small as 1μL
- UV, blue and white LED excitation, covering a broad wavelength range
- No filter changes and no expensive monochromator
- Ability to run diverse and customizable applications
- Ability to measure multiple fluorophores from a single sample
- Spectral and quantitative data