Anti-ITGAX Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: N418]

Lieferant: Tonbo Biosciences
20-0114-U100 20-0114-U025
20-0114-U100EA 113 EUR
20-0114-U100 20-0114-U025
Anti-ITGAX Armenian Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: N418]
Antikörper
The N418 antibody reacts with mouse CD11c, also known as integrin alpha X. This 150 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18) into a receptor complex known as CR4 which can bind and induce signaling through ICAMs and VCAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.

The N418 antibody is widely used as a marker for CD11c expression on dendritic cells (DC), often in parallel with markers for CD11b, for identification of developmental stages and mature subsets of this cell type. CD11c is prominently expressed on tissue macrophages, and is also detected on some types of activated T cells and intestinal intraepithelial lymphocytes (IEL).

Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: monoclonal
Clone: N418
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR