Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1D3]

Lieferant: Tonbo Biosciences
20-0193-U025 20-0193-U100
20-0193-U025EA 53.3 EUR
20-0193-U025 20-0193-U100
Anti-CD19 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 1D3]
Antikörper
The 1D3 antibody reacts with mouse CD19, a 95 kDa glycoprotein which acts as a co-receptor, along with CD21 and CD81, in support of the functional B cell receptor (BCR). This complex provides antigen-specific recognition and subsequent activation of B cells to proliferate and differentiate into antibody-secreting cells (plasma cells) or memory B cells, which are crucial for secondary antigen encounter. CD19 is a lineage-differentiation marker, as its expression is detectable at the earliest B cell stages, through development, and is finally lost upon transition to mature plasma cells.

The 1D3 antibody is widely used as a phenotypic marker for CD19 expression on B cells, as well as on dendritic cell subsets.

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: monoclonal
Clone: 1D3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR