Rat IgG1 Isotype Control [clone: HRPN] (APC (Allophycocyanin))

Lieferant: Tonbo Biosciences
20-4301-U100
20-4301-U100EA 110 EUR
20-4301-U100
Rat IgG1 Isotype Control [clone: HRPN] (APC (Allophycocyanin))
Antikörper
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: IgG2b (Rat) (rat IgG2b)
Clonality: monoclonal
Clone: HRPN
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR