Anti-PTPRC Rat Antibody (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]

Lieferant: Tonbo Biosciences
25-4031-U100
25-4031-U100EA 108 EUR
25-4031-U100
Anti-PTPRC Rat Antibody (APC (Allophycocyanin)-Cy7®) [clone: LTF-2]
Antikörper
Type: Control
Antigen: PTPRC
Clonality:
Clone: LTF-2
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: 0
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR