Anti-IgE Goat Polyclonal Antibody (Biotin)

Lieferant: Tonbo Biosciences
30-8026-M001
30-8026-M001EA 238 EUR
30-8026-M001
Anti-IgE Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Human IgE (epsilon chain) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: ε chain
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR