Anti-IgM Goat Polyclonal Antibody (Biotin)
Lieferant: Tonbo Biosciences
30-8030-M001
30-8030-M001EA 136 EUR
30-8030-M001
Anti-IgM Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Human IgM (mu chain) - Affinity Pure, Biotin Conjugate, min x w/human IgA + IgG
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human