Anti-IgM Goat Polyclonal Antibody (Biotin)

Lieferant: Tonbo Biosciences
30-8065-M002
30-8065-M002EA 251 EUR
30-8065-M002
Anti-IgM Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Mouse IgM (mu chain) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR