Anti-IgG Goat Polyclonal Antibody (Biotin)

Lieferant: Tonbo Biosciences
30-8067-M002
30-8067-M002EA 124 EUR
30-8067-M002
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antikörper
Goat anti-Rabbit IgG (H&L) - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR