Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Tonbo Biosciences
35-8016-M001
35-8016-M001EA 158 EUR
35-8016-M001
Anti-IgA Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Goat anti-Human IgA (alpha chain) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: α chain
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR