Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Tonbo Biosciences
35-8030-M001
35-8030-M001EA 124 EUR
35-8030-M001
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Goat anti-Human IgM (mu chain) - Affinity Pure, FITC Conjugate, min x w/human IgA + IgG

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR