Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Tonbo Biosciences
35-8057-U500
35-8057-U500EA 259 EUR
35-8057-U500
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Goat anti-Mouse IgG (H&L) - F(ab')2 fragment, FITC Conjugate, min x w/bovine, goat, human, rabbit, or rat IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR