Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Tonbo Biosciences
35-8070-M002
35-8070-M002EA 165 EUR
35-8070-M002
Anti-IgG Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Goat anti-Rabbit IgG (H&L) - Affinity Pure, FITC Conjugate, min x w/bovine, human, or mouse IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR