Rat IgG1 Isotype Control [clone: HRPN]
Lieferant: Tonbo Biosciences
40-4301-U100
40-4301-U100EA 78.9 EUR
40-4301-U100
Rat IgG1 Isotype Control [clone: HRPN]
Antikörper
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.