Anti-FCER1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MAR-1]

Lieferant: Tonbo Biosciences
50-5898-U025 50-5898-U100
50-5898-U025EA 46.9 EUR
50-5898-U025 50-5898-U100
Anti-FCER1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: MAR-1]
Antikörper
The MAR-1 antibody reacts with the Fc epsilon Receptor I alpha chain (FceRIa), a transmembrane protein member of the Ig superfamily. This chain, together with a beta chain and two gamma chains form a tetrameric complex that supports IgE-mediated signaling and subsequent release of chemical mediators of allergy and immediate hypersensitivity. FceR1a is upregulated in the presence of IgE on those cell types which express it, such as Mast cells and Basophils.

The MAR-1 antibody is widely used both in flow cytometry and for depletion of cells in vitro / in vivo. Please choose the appropriate format for each application.

Type: Primary
Antigen: FCER1 (Fc fragment of IgE, high affinity I, receptor for)
Clonality: monoclonal
Clone: 42064
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR