Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: Tonbo Biosciences
50-8034-M001
50-8034-M001EA 408 EUR
50-8034-M001
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Goat anti-mouse IgG (H+L), secondary antibody, conjugated to SureLight® R-Phycoerythrin, 1mg
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse