Anti-IgG2b Goat Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: Tonbo Biosciences
50-8035-U100
50-8035-U100EA 273 EUR
50-8035-U100
Anti-IgG2b Goat Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Goat anti-Mouse IgG2b, secondary antibody, conjugated to SureLight® R-Phycoerythrin

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR