Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: Tonbo Biosciences
50-8036-M001
50-8036-M001EA 408 EUR
50-8036-M001
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Goat anti-Rabbit IgG, secondary antibody, conjugated toSureLight® R-Phycoerythrin

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR