Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: HI100]

Lieferant: Tonbo Biosciences
60-0458-T100 60-0458-T025
60-0458-T100EA 199 EUR
60-0458-T100 60-0458-T025
Anti-CD45RA Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: HI100]
Antikörper
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB and CD45RO.

The HI100 antibody is widely used as a marker for human CD45RA expression on naïve and activated T cells, B cells, and monocytes.

Type: Primary
Antigen: CD45RA (CD45RA)
Clonality: monoclonal
Clone: HI100
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR