Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy7®)
Lieferant: Tonbo Biosciences
60-4031-U100
60-4031-U100EA 108 EUR
60-4031-U100
Rat IgG2b Isotype Control [clone: LTF-2] (PE (Phycoerythrin)-Cy7®)
Antikörper
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.