Mouse IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin)/Cy7®)
Lieferant: Tonbo Biosciences
60-4714-U100
60-4714-U100EA 108 EUR
60-4714-U100
Mouse IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin)/Cy7®)
Antikörper
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen