Rat IgG2b Isotype Control [clone: LTF-2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Lieferant: Tonbo Biosciences
65-4031-U100
65-4031-U100EA 110 EUR
65-4031-U100
Rat IgG2b Isotype Control [clone: LTF-2] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antikörper
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: IgG2b (Rat) (rat IgG2b)
Clonality: monoclonal
Clone: LTF-2
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR