Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)

Lieferant: Tonbo Biosciences
65-4714-U100
65-4714-U100EA 135 EUR
65-4714-U100
Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Antikörper
PerCP-Cyanine5.5 Mouse IgG1 Isotype Control (MOPC-21)

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PerCP-Cy5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR