Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Lieferant: Tonbo Biosciences
65-4724-U100
65-4724-U100EA 117 EUR
65-4724-U100
Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Antikörper
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: C1.18.4
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR