Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex))
Lieferant: Tonbo Biosciences
67-4724-U100
67-4724-U100EA 122 EUR
67-4724-U100
Mouse IgG2a Isotype Control [clone: C1.18.4] (PerCP (Peridinin-Chlorophyll Protein Complex))
Antikörper
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: control
Antigen: Isotype Control
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen