Anti-ITGAM Mouse Monoclonal Antibody [clone: ICRF44]
Lieferant: Tonbo Biosciences
70-0118-U100
70-0118-U100EA 46.9 EUR
70-0118-U100
Anti-ITGAM Mouse Monoclonal Antibody [clone: ICRF44]
Antikörper
The ICRF44 antibody reacts with human CD11b, also known as integrin alpha M. This 165-170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles with Integrin beta-2More Product Information