Rat IgG2b Isotype Control [clone: LTF-2]

Lieferant: Tonbo Biosciences
70-4031-U100
70-4031-U100EA 53.3 EUR
70-4031-U100
Rat IgG2b Isotype Control [clone: LTF-2]
Antikörper
The LTF-2 immunoglobulin is useful as an isotype-matched control. The LTF-2 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2b antibodies.

Type: Control
Antigen: LTF-2
Clonality:
Clone: LTF-2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: N/A
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR