Rat IgG2a kappa Isotype Control [clone: 2A3]

Lieferant: Tonbo Biosciences
70-4321-U100
70-4321-U100EA 53.3 EUR
70-4321-U100
Rat IgG2a kappa Isotype Control [clone: 2A3]
Antikörper
Purified Rat IgG2a Isotype Control (2A3)

Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR