Mouse IgG2a Isotype Control [clone: C1.18.4]
Lieferant: Tonbo Biosciences
70-4724-U100
70-4724-U100EA 59.7 EUR
70-4724-U100
Mouse IgG2a Isotype Control [clone: C1.18.4]
Antikörper
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: C1.18.4
Clonality:
Clone: C1.18.4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: N/A