Anti-IgM Goat Polyclonal Antibody
Lieferant: Tonbo Biosciences
70-8028-M002
70-8028-M002EA 136 EUR
70-8028-M002
Anti-IgM Goat Polyclonal Antibody
Antikörper
Goat anti-Human IgM (mu chain) - Affinity Pure
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human