Anti-IgG Goat Polyclonal Antibody
Lieferant: Tonbo Biosciences
70-8042-M002
70-8042-M002EA 109 EUR
70-8042-M002
Anti-IgG Goat Polyclonal Antibody
Antikörper
Goat anti-Mouse IgG (H&L) - Affinity Pure
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse