Anti-IgM (µ chain) Rabbit Polyclonal Antibody
Lieferant: Tonbo Biosciences
70-8075-M001
70-8075-M001EA 195 EUR
70-8075-M001
Anti-IgM (µ chain) Rabbit Polyclonal Antibody
Antikörper
Rabbit anti-Human IgM (mu chain) - Affinity Pure
Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human