Anti-IgG Rabbit Polyclonal Antibody

Lieferant: Tonbo Biosciences
70-8077-M002
70-8077-M002EA 165 EUR
70-8077-M002
Anti-IgG Rabbit Polyclonal Antibody
Antikörper
Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/human serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR