Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Tonbo Biosciences
72-8076-M001
72-8076-M001EA 147 EUR
72-8076-M001
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Rabbit anti-Mouse IgG (H&L) - Affinity Pure, HRP Conjugate
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: Mouse