Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Tonbo Biosciences
72-8077-M001
72-8077-M001EA 158 EUR
72-8077-M001
Anti-IgG Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Rabbit anti-Mouse IgG (H&L) - Affinity Pure, HRP Conjugate, min x w/human serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR