Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Tonbo Biosciences
72-8084-M001
72-8084-M001EA 251 EUR
72-8084-M001
Anti-IgG Donkey Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Donkey anti-Rabbit IgG (H&L) - Affinity Pure, HRP Conjugate, min x w/bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat or sheep

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR