Rat IgG1 Isotype Control [clone: HRPN] (violetFluor® 450)

Lieferant: Tonbo Biosciences
75-4301-U100
75-4301-U100EA 108 EUR
75-4301-U100
Rat IgG1 Isotype Control [clone: HRPN] (violetFluor® 450)
Antikörper
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.

Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation: violetFluor™ 450
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: N/A
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR