SilverOUT™, G-Biosciences
Lieferant: G Bioscienes
786-244
786-244EA
340
EUR
786-244
SilverOUT™, G-Biosciences
Protein-Extraktion
Proteinextraktions-Reagenzien
SilverOUT removes interfering silver ions from gel pieces.
Gel spots can be destained within 10 minutes.
Compatible with downstream processes including mass spectrometry.