Immobilised Ficin
Lieferant: G Bioscienes
786-793
786-793EA
528
EUR
786-793
Immobilised Ficin
Protein-Extraktion
Proteinextraktions-Reagenzien
Ficin (or Ficain) is a cysteine protease enzyme (EC 3.4.22.3) isolated from fig latex is that has the endopeptidase activity to cleave Immunoglobulin G (IgG) molecules in the hinge region.
- Generate Fab and F(ab‘)₂ fragments
- For digestion of mouse IgG₁
- Eliminates contamination by Ficin