Anti-TER-119 Rat Monoclonal Antibody (redFluor™ 710) [clone: TER-119]

Lieferant: Tonbo Biosciences
80-5921-U025 80-5921-U100
80-5921-U025EA 59.7 EUR
80-5921-U025 80-5921-U100
Anti-TER-119 Rat Monoclonal Antibody (redFluor™ 710) [clone: TER-119]
Antikörper
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.

Type: Primary
Antigen: TER-119
Clonality: Monoclonal
Clone: TER-119
Conjugation: redFluor™ 710
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR