Anti-HLA-DR Mouse Monoclonal Antibody (redFluor™ 710) [clone: L243]

Lieferant: Tonbo Biosciences
80-9952-T025 80-9952-T100
80-9952-T025EA 109 EUR
80-9952-T025 80-9952-T100
Anti-HLA-DR Mouse Monoclonal Antibody (redFluor™ 710) [clone: L243]
Antikörper
redFluor™ 710 Anti-Human HLA-DR (L243)

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: L243
Conjugation: redFluor™ 710
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Chimpanzee, Cynomolgus, Rhesus, Baboon, Squirrel Monkey, African Green Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR