Anti-CD3, CD11b, CD45/B220, Ly-6G, TER-119 Rat Monoclonal Antibody (FITC) [clone: 17A2, M1/70, RA3-6B2, RB6-8C5, TER-119]

Lieferant: Tonbo Biosciences
92-7770-T100
92-7770-T100EA 369 EUR
92-7770-T100
Anti-CD3, CD11b, CD45/B220, Ly-6G, TER-119 Rat Monoclonal Antibody (FITC) [clone: 17A2, M1/70, RA3-6B2, RB6-8C5, TER-119]
Antikörper
The FITC Mouse Lineage Cocktail contains a combination of pre-diluted antibodies that identify lineage committed cells by flow cytometry and facilitate their exclusion from hematopoietic progenitor cells in mouse bone marrow. The antibodies in the cocktail react with cells from major hematopoietic lineages including T, B and NK cells, monocytes/macrophages, granulocytes and erythrocytes. The Lineage Cocktail contains the following FITC conjugated antibodies: anti-mouse CD3 (17A2), anti-mouse CD11b (M1/70), anti-mouse CD45/B220 (RA3-6B2), anti-mouse Ly-6G (Gr-1) (RB6-8C5) and anti-mouse TER-119 (TER-119). An Isotype Control Cocktail consisting of FITC conjugated isotype-matched control antibodies (Rat IgG2a, kappa; Rat IgG2b, kappa) is also provided in a separate vial.

Type: Primary
Antigen: CD3, CD11b, CD45/B220, Ly-6G, TER-119
Clonality: Monoclonal
Clone: 17A2, M1/70, RA3-6B2, RB6-8C5, TER-119
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa/IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR