Experimentation kits, protein electrophoresis
Lieferant: G-Biosciences
BE-406
BE-406EA 329 EUR
BE-406
Experimentation kits, protein electrophoresis
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
Using the Protein Electrophoresis kit, students will learn the principles of various types of electrophoresis, including denaturing and non-denaturing electrophoresis, and how this powerful technique is used to analyse proteins. The kit will introduce students to the different separation matrices currently in use and will understand their differing separation properties and their role in protein analysis. Students have an option of casting their own electrophoresis gels using polyacrylamide or using pre-cast commercially available gels. This kit is provided with all of the reagents, buffers and supplies needed for casting acrylamide gels, preparing protein samples, running electrophoresis, and staining the gels for visualisation of protein bands. Test protein samples and protein standards are also provided.
  • Principle of protein electrophoresis and its various applications
  • Hands-on lab activities to run electrophoresis
  • Casting of electrophoresis gels, protein sample preparation, and running electrophoresis with test samples
  • Staining electrophoresis gels for visualization of resolved protein bands
  • Use of protein standard for determination of protein mass
Also required: Protein electrophoresis equipment, water bath or beaker and thermometer.
Lieferumfang: Supplied with components needed for hands-on experimentation for six workstations of 4 to 5 students or 24 to 30 students. Supplied with Teacher’s Guide and separate Student’s Guides.