Beauchene adult human skull model
Lieferant: Binhold, Paul Lehrmittel
1000069
BINH1000069EA 0 EUR
BINH1000069
Beauchene adult human skull model
Modelle Humanbiologie
Beauchene adult human skull is a natural cast and makes the complexity of the anatomical structure easy to understand, since it is disassembled into its 22 individual bones.
  • Bones/bone pairs are coloured to make dentification easier
  • Bones can be reassembled by means of inconspicuous, stable connectors attached at the slightly simplified skull sutures
  • Human skull consists of individual bones that gradually grow together as the development proceeds
This skull model is a great tool for teaching and learning the complex anatomy of the human skull.