Experimentation kits, DNA/RNA microarrays
Lieferant: EDVOTEK
235
EDVO235EA 149 EUR
EDVO235
Experimentation kits, DNA/RNA microarrays
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
Microarray technology allows scientists to screen large numbers of samples in one assay. This technology allows high-throughput screening of small samples and allows comparisons to be made between healthy and unhealthy tissues. In this experiment, students apply simulated DNA and RNA samples to a membrane to screen for positive and negative samples.
  • For 10 lab groups
  • Complete in 60 minutes
Also required: 5 µl fixed volume or adjustable micropipette and tips.
Lieferumfang: Kit includes instructions, simulated patient DNA and RNA samples and controls, microarray cards, plastic bags to incubate membrane, microtest tubes and pipettes.