Anti-OPN Mouse Monoclonal Antibody [clone: 87-B]

Lieferant: ENZO LIFE SCIENCES
ADI905630100
ENZOADI905630100EA 422 EUR
ENZOADI905630100
Anti-OPN Mouse Monoclonal Antibody [clone: 87-B]
Antikörper
Osteopontin (OPN) is an acidic extracellular matrix cell adhesion protein that is relatively abundant not only in bone matrix, plasma, urine, and milk, but is also found in malignant and atherogenic tissues. Phosphorylation, glycosylation and calcium modifications allow intact and fragmented Osteopontin to direct a variety of diverse responses including tissue remodeling, inflammation and cell survival. Plasma Osteopontin has been shown to provide prognostic information in breast, prostate, colon, and lung cancers as well as metastatic carcinomas. The notable presence of Osteopontin in a variety of tumors is strongly correlated to pathological stage, suggesting its critical role in tumor invasiveness, progression and metastasis. In addition, Osteopontin inhibits inducible nitric oxide synthase activity, thereby protecting tumor cells from NO-mediated macrophage cytotoxic attack. Osteopontin is found in atherosclerotic plaques and may drive a number of diabetic vascular pathologies.

Type: Primary
Antigen: OPN
Clonality: Monoclonal
Clone: 87-B
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR