Anti-APP Mouse Monoclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADI905804100
ENZOADI905804100EA 533 EUR
ENZOADI905804100
Anti-APP Mouse Monoclonal Antibody
Antikörper
The β-amyloid peptide (β A4), proteolytically released from the amyloid precursor protein (APP), provides the principal component of senile plaques in Alzheimer's disease. Cleavage of APP by α-secretase or alternatively by β-secretase leads to generation and extracellular release of soluble APP peptides, S-APP-α and S-APP-β, respectively, and the retention of corresponding membrane-anchored C-terminal fragments, C83 and C99. Subsequent processing of C83 by γ-secretase yields P3 peptides. This is the major secretory pathway and is nonamyloidogenic. Alternatively, presenilin/ nicastrin-mediated γ-secretase processing of C99 releases the amyloid β proteins, amyloid-β 40 (Aβ40) and amyloid-β 42 (Aβ42), major components of amyloid plaques, and the cytotoxic C-terminal fragments, γ-CTF(50), γ-CTF(57) and γ-CTF(59).

  • For ELISA, Flow Cytometry, Immunocytochemistry, Western Blot

Recognizes human amyloid beta A4 (CT, 1-42). Does not cross react with beta A4 (1-40). Detects a band of ~4kDa by Western blot.

Reconstitute with 1ml water (15 minutes at room temperature).

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR