Anti-PGF2 alpha Receptor Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADI905901100
ENZOADI905901100EA 500 EUR
ENZOADI905901100
Anti-PGF2 alpha Receptor Rabbit Polyclonal Antibody
Antikörper
Arachidonic acid is converted by cyclooxygenase (COX) to prostaglandin H2 (PGH2), which is further converted to the derivative prostanoids PGD2, PGE2, PGF2α, PGI2 (prostacyclin) and thromboxane A2 (TXA2). Five classes of prostanoid receptors are recognized and named DP, EP, FP, IP and TP receptors corresponding to the agonist properties of the associated prostanoids. FP receptors are found in smooth muscle where they mediate contraction, and are believed to be coupled via Gq/11 subunits to stimulate phosphatidyl inositol (PI) hydrolysis. The FP agonist Fluprostenol (a PGF2α analogue) is used in animal husbandry to synchronize oestrus and induce partruition. FP receptors are known to mediate intraocular pressure, and FP agonists such as latanoprost and travoprost are drugs commonly use to treat glaucoma.

Recognizes mouse and rat PGF2 alpha Receptor (FP). Predicted MW of ~41kDa, observed by Western blot ~45kDa. Higher MW species (~60kDa) and intermediates also observed by Western blot which reflect post-translational modification of the receptor. Predicted species reactivity based on epitope sequence homology: wild boar, dog.

For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Bestellinformation: Manufactured by Proteimax.

Type: Primary
Antigen: PGF2 ALPHA RECEPTOR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR