Anti-TAP1 Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADICSA620E
ENZOADICSA620EEA 437 EUR
ENZOADICSA620E
Anti-TAP1 Rabbit Polyclonal Antibody
Antikörper
TAP1 and TAP2 are two structurally related subunits of the Transporter associated with antigen processing (TAP). The TAP complex is a member of the ATP binding cassette (ABC) family of transmembrane transporters. The TAP transporter is an essential component of the MHC class I antigen presentation pathway by binding peptides in its cytosolic part and subsequently translocating the peptides into the lumen of the endoplasmic reticulum where assembly of MHC class I and pepide takes place.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and rat TAP1. Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: TAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR