Anti-BVR Rabbit Polyclonal Antibody

Lieferant: ENZO LIFE SCIENCES
ADIOSA450E
ENZOADIOSA450EEA 482 EUR
ENZOADIOSA450E
Anti-BVR Rabbit Polyclonal Antibody
Antikörper
The phosphoprotein BVR (Biliverdin Reductase) is an oxireductase which catalyzes the reduction of the gamma-meso bridge of Biliverdin to Bilirubin. BVR is unique among all enzymes characterized to date because BVR has dual pH/dual cofactor requirements. The reductase is highly conserved in its primary structure and molecular properties and uses NADH in the acidic range (peak activity at pH~6.7), whereas NADPH is utilized in the basic range (peak activity at pH ~8.7).

  • For Immunoprecipitation, Western Blot

Recognizes rat, mouse and bacteria (E. coli) Biliverdin Reductase. Detects bands of ~33kDa by Western blot.

Type: Primary
Antigen: BVR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR